Ethyl 2-fluoro-6-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

34988-1G 34988-25G 34988-500MG 34988-5G
76941-076EA 248.26 USD
76941-076 76941-078 76941-080 76941-082
Ethyl 2-fluoro-6-(trifluoromethyl)benzoate ≥97%
Ethyl 2-fluoro-6-(trifluoromethyl)benzoate

Precaución: for Research Use Only

Formula: C₁₀H₈F₄O₂
MW: 236.163 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06204410
CAS Number: 773134-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR