5-tert-Butyl-2-chlorophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

34952-1G 34952-25G 34952-500MG 34952-5G
76941-060EA 213.25 USD
76941-060 76941-062 76941-064 76941-066
5-tert-Butyl-2-chlorophenylboronic acid ≥97%
5-tert-Butyl-2-chlorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₄BClO₂
Storage Temperature: Refrigerator
CAS Number: 1233056-75-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR