Methyl 5-bromo-2-fluoro-3-methylbenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
34923-1G
34923-25G
34923-5G
76941-048EA
88.06
USD
76941-048
76941-050
76941-052
Methyl 5-bromo-2-fluoro-3-methylbenzoate ≥97%
Methyl 5-bromo-2-fluoro-3-methylbenzoate
Precaución: for Research Use Only
Formula:
C₉H₈BrFO₂ MW: 247.061 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD24115508 CAS Number: 1538333-27-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...