Methyl 6-bromo-2-chloro-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

34917-1G 34917-25G 34917-500MG 34917-5G
76941-032EA 266.29 USD
76941-032 76941-034 76941-036 76941-038
Methyl 6-bromo-2-chloro-3-methylbenzoate ≥97%
Methyl 6-bromo-2-chloro-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD31805777
CAS Number: 2244107-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR