6-Bromo-2-fluoro-3-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

34883-1G 34883-500MG 34883-5G
76941-018EA 471.04 USD
76941-018 76941-020 76941-022
6-Bromo-2-fluoro-3-methylpyridine ≥97%
6-Bromo-2-fluoro-3-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFN
Storage Temperature: Ambient
MDL Number: MFCD18254091
CAS Number: 1211536-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR