Methyl 3,4-difluoro-2-hydroxybenzoate ≥97%

Supplier: AOBChem USA

Warning

34768-1G 34768-500MG 34768-5G
76940-976EA 561.22 USD
76940-976 76940-978 76940-980
Methyl 3,4-difluoro-2-hydroxybenzoate ≥97%
Methyl 3,4-difluoro-2-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₆F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD11042797
CAS Number: 1214362-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR