4-Bromo-5-thiazolecarboxylic acid ethyl ester ≥97%

Supplier: AOBChem USA

Warning

34702-1G 34702-25G 34702-500MG 34702-5G
76940-948EA 142.17 USD
76940-948 76940-950 76940-952 76940-954
4-Bromo-5-thiazolecarboxylic acid ethyl ester ≥97%
4-Bromo-5-thiazolecarboxylic acid ethyl ester
Formula: C₆H₆BrNO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD09909677
CAS Number: 152300-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR