6-(4-Formylphenyl)nicotinaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-(4-Formylphenyl)pyridine-3-carbaldehyde

34681-1G 34681-25G 34681-500MG 34681-5G
76940-934EA 385.11 USD
76940-934 76940-936 76940-938 76940-940
6-(4-Formylphenyl)nicotinaldehyde ≥97%
6-(4-Formylphenyl)nicotinaldehyde
Formula: C₁₃H₉NO₂
MW: 211.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659608
CAS Number: 885950-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR