Methyl 2-bromo-6-chloro-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

34620-1G 34620-25G 34620-500MG 34620-5G
76940-906EA 183.54 USD
76940-906 76940-908 76940-910 76940-912
Methyl 2-bromo-6-chloro-3-methylbenzoate ≥97%
Methyl 2-bromo-6-chloro-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrClO₂
MW: 263.516 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619516
CAS Number: 2092387-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR