Ethyl 2-chloro-5-methylbenzoate ≥95%

Supplier: AOBChem USA

Warning

34614-1G 34614-25G 34614-5G
76940-892EA 99.73 USD
76940-892 76940-894 76940-896
Ethyl 2-chloro-5-methylbenzoate ≥95%
Ethyl 2-chloro-5-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
MW: 198.646 g/mol
Storage Temperature: Ambient
CAS Number: 945262-03-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR