Methyl 2,3-difluoro-4-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

34606-1G 34606-25G 34606-5G
76940-880EA 107.16 USD
76940-880 76940-882 76940-884
Methyl 2,3-difluoro-4-methoxybenzoate ≥97%
Methyl 2,3-difluoro-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈F₂O₃
Storage Temperature: Refrigerator
MDL Number: MFCD11520389
CAS Number: 544704-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR