Methyl 4'-fluoro-1,1'-biphenyl-4-carboxylate ≥97%

Supplier: AOBChem USA

Warning

34590-1G 34590-25G 34590-500MG 34590-5G
76940-850EA 257.8 USD
76940-850 76940-852 76940-854 76940-856
Methyl 4'-fluoro-1,1'-biphenyl-4-carboxylate ≥97%
Methyl 4'-fluoro-1,1'-biphenyl-4-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₁FO₂
MW: 230.234 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05979321
CAS Number: 80254-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR