Methyl 3-Chloro-5-(Methylsulfanyl)Benzoate ≥95%

Supplier: AOBChem USA

Warning

34399-1G 34399-25G 34399-500MG 34399-5G
76940-772EA 240.83 USD
76940-772 76940-774 76940-776 76940-778
Methyl 3-Chloro-5-(Methylsulfanyl)Benzoate ≥95%
Methyl 3-Chloro-5-(Methylsulfanyl)Benzoate

Precaución: for Research Use Only

Formula: C₉H₉ClO₂S
MW: 216.685 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22488876
CAS Number: 1824578-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR