Methyl 2,3-Dichloro-6-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

34372-1G 34372-500MG 34372-5G
76940-760EA 448.77 USD
76940-760 76940-762 76940-764
Methyl 2,3-Dichloro-6-methoxybenzoate ≥97%
Methyl 2,3-Dichloro-6-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈Cl₂O₃
MW: 235.064 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31619483
CAS Number: 2179038-35-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR