Ethyl 3,4,5-trichlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

34343-1G 34343-25G 34343-500MG 34343-5G
76940-744EA 266.29 USD
76940-744 76940-746 76940-748 76940-750
Ethyl 3,4,5-trichlorobenzoate ≥97%
Ethyl 3,4,5-trichlorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇Cl₃O₂
MW: 253.51 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14704256
CAS Number: 120530-42-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR