Ethyl 3-chloro-2-fluoro-5-(trifluoromethyl)benzoate ≥95%
Supplier: AOBChem USA
|
Warning
|
34246-1G
34246-25G
34246-500MG
34246-5G
76940-700EA
183.54
USD
76940-700
76940-702
76940-704
76940-706
Ethyl 3-chloro-2-fluoro-5-(trifluoromethyl)benzoate ≥95%
Ethyl 3-chloro-2-fluoro-5-(trifluoromethyl)benzoate
Precaución: for Research Use Only
Formula:
C₁₀H₇ClF₄O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD06204522 CAS Number: 773135-53-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...