Ethyl 2,4-dichloro-3-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

34237-1G 34237-25G 34237-500MG 34237-5G
76940-686EA 171.87 USD
76940-686 76940-688 76940-690 76940-692
Ethyl 2,4-dichloro-3-fluorobenzoate ≥97%
Ethyl 2,4-dichloro-3-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₇Cl₂FO₂
MW: 237.055 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786000
CAS Number: 1803718-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR