Ethyl 2,4-difluoro-5-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

34233-1G 34233-500MG 34233-5G
76940-672EA 683.22 USD
76940-672 76940-674 76940-676
Ethyl 2,4-difluoro-5-methoxybenzoate ≥97%
Ethyl 2,4-difluoro-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD28789271
CAS Number: 1803729-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR