5-Phenylnicotinic acid ≥97%

Supplier: AOBChem USA

Warning

34141-1G 34141-25G 34141-5G
76940-658EA 99.73 USD
76940-658 76940-660 76940-662
5-Phenylnicotinic acid ≥97%
5-Phenylnicotinic acid
Formula: C₁₂H₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD03086176
CAS Number: 10177-12-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR