N-Boc-5-bromo-2-methylaniline ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: t-Butyl 5-bromo-2-methylphenylcarbamate

34128-25G 34128-5G
76940-650EA 310.85 USD
76940-650 76940-652
N-Boc-5-bromo-2-methylaniline ≥97% research grade
N-Boc-5-bromo-2-methylaniline
Formula: C₁₂H₁₆BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD11504830
CAS Number: 221538-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR