6-Bromo-7-chloro-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

34121-1G 34121-500MG 34121-5G
76940-636EA 683.22 USD
76940-636 76940-638 76940-640
6-Bromo-7-chloro-1H-indazole ≥97%
6-Bromo-7-chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.477 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23705449
CAS Number: 1427405-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR