Methyl 3-bromo-4-chloro-5-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

34111-25G 34111-5G
76940-626EA 495.45 USD
76940-626 76940-628
Methyl 3-bromo-4-chloro-5-fluorobenzoate ≥97%
Methyl 3-bromo-4-chloro-5-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD11846086
CAS Number: 1160574-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR