Ethyl 2,4-dimethylthiophene-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

33831-1G 33831-500MG 33831-5G
76940-588EA 635.48 USD
76940-588 76940-590 76940-592
Ethyl 2,4-dimethylthiophene-3-carboxylate ≥97%
Ethyl 2,4-dimethylthiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₂O₂S
MW: 184.255 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30294241
CAS Number: 36382-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR