Methyl 3,6-dichloro-2-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

33756-1G 33756-25G 33756-5G
76940-578EA 64.72 USD
76940-578 76940-580 76940-582
Methyl 3,6-dichloro-2-fluorobenzoate ≥97%
Methyl 3,6-dichloro-2-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₅Cl₂FO₂
MW: 223.029 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698030
CAS Number: 1214324-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR