5-Isoquinolinecarboxylic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Isoquinoline-5-carboxylic acid

33675-25G 33675-5G
76940-552EA 424.36 USD
76940-552 76940-554
5-Isoquinolinecarboxylic acid ≥97%
5-Isoquinolinecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₂
MW: 173.17 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03788744
CAS Number: 27810-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR