Dimethyl 3-bromo-4-chlorophthalate ≥97%

Supplier: AOBChem USA

Warning

33660-1G 33660-500MG 33660-5G
76940-538EA 683.22 USD
76940-538 76940-540 76940-542
Dimethyl 3-bromo-4-chlorophthalate ≥97%
Dimethyl 3-bromo-4-chlorophthalate

Precaución: for Research Use Only

Formula: C₁₀H₈BrClO₄
MW: 307.525 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31561311
CAS Number: 1855005-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR