Methyl 3-(benzyloxy)-5-bromobenzoate ≥97%

Supplier: AOBChem USA

Warning

33600-1G 33600-25G 33600-500MG 33600-5G
76940-524EA 272.66 USD
76940-524 76940-526 76940-528 76940-530
Methyl 3-(benzyloxy)-5-bromobenzoate ≥97%
Methyl 3-(benzyloxy)-5-bromobenzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₃BrO₃
MW: 321.166 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27578340
CAS Number: 1820683-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR