2-Benzylamino-5-bromopyrimidine ≥97%

Supplier: AOBChem USA

Warning

33564-1G 33564-25G 33564-5G
76940-512EA 64.72 USD
76940-512 76940-514 76940-516
2-Benzylamino-5-bromopyrimidine ≥97%
2-Benzylamino-5-bromopyrimidine

Precaución: for Research Use Only

Formula: C₁₁H₁₀BrN₃
Storage Temperature: Ambient
MDL Number: MFCD00483264
CAS Number: 38373-55-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR