Benzyl 2-bromo-6-methoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

33532-1G 33532-25G 33532-5G
76940-500EA 62.6 USD
76940-500 76940-502 76940-504
Benzyl 2-bromo-6-methoxybenzoate ≥95%
Benzyl 2-bromo-6-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₅H₁₃BrO₃
Storage Temperature: Ambient
MDL Number: MFCD30834074
CAS Number: 2121512-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR