5-Methyl-1H-indazole-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

33495-1G 33495-25G 33495-500MG 33495-5G
76940-470EA 260.99 USD
76940-470 76940-472 76940-474 76940-476
5-Methyl-1H-indazole-4-carbaldehyde ≥97%
5-Methyl-1H-indazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₈N₂O
MW: 160.173 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28516466
CAS Number: 1783447-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR