Methyl 4-chloro-3-(trifluoromethoxy)benzoate ≥97%

Supplier: AOBChem USA

Warning

33380-1G 33380-25G 33380-500MG 33380-5G
76940-444EA 198.39 USD
76940-444 76940-446 76940-448 76940-450
Methyl 4-chloro-3-(trifluoromethoxy)benzoate ≥97%
Methyl 4-chloro-3-(trifluoromethoxy)benzoate

Precaución: for Research Use Only

Formula: C₉H₆ClF₃O₃
MW: 254.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18399027
CAS Number: 1261444-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR