9-(3,5-Dibromophenyl)carbazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 9-(3,5-Dibromophenyl)-9H-carbazole

33317-25G 33317-5G
76940-432EA 247.19 USD
76940-432 76940-434
9-(3,5-Dibromophenyl)carbazole ≥97%
9-(3,5-Dibromophenyl)carbazole

Precaución: for Research Use Only

Formula: C₁₈H₁₁Br₂N
MW: 401.10 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD27939624
CAS Number: 750573-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR