5-Iodo-2-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

33243-25G 33243-5G
76940-416EA 524.09 USD
76940-416 76940-418
5-Iodo-2-methoxybenzaldehyde ≥97%
5-Iodo-2-methoxybenzaldehyde
Formula: C₈H₇IO₂
Storage Temperature: Refrigerator
MDL Number: MFCD04128999
CAS Number: 42298-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR