Benzyl 3-fluoro-5-methylbenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
33023-25G
33023-5G
76940-400EA
310.85
USD
76940-400
76940-402
Benzyl 3-fluoro-5-methylbenzoate ≥97%
Benzyl 3-fluoro-5-methylbenzoate
Precaución: for Research Use Only
Formula:
C₁₅H₁₃FO₂ MW: 244.261 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD30537120 CAS Number: 2070896-26-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...