Methyl 4-formyl-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

33006-1G 33006-25G 33006-5G
76940-376EA 114.58 USD
76940-376 76940-378 76940-380
Methyl 4-formyl-3-methylbenzoate ≥97%
Methyl 4-formyl-3-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
Storage Temperature: Ambient
MDL Number: MFCD20923636
CAS Number: 24078-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR