Methyl 6-fluorobenzo[b]thiophene-2-carboxylate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Fluoro-benzo[b]thiophene-2-carboxylic acid methyl ester

32881-1G 32881-25G 32881-5G
76940-354EA 63.66 USD
76940-354 76940-356 76940-358
Methyl 6-fluorobenzo[b]thiophene-2-carboxylate ≥97%
Methyl 6-fluorobenzo[b]thiophene-2-carboxylate
Formula: C₁₀H₇FO₂S
MW: 210.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11044557
CAS Number: 550998-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR