Methyl 2,6-difluoro-3-methoxybenzoate ≥95%
Supplier: AOBChem USA
|
Warning
|
32859-1G
32859-25G
32859-500MG
32859-5G
76940-338EA
153.84
USD
76940-338
76940-340
76940-342
76940-344
Methyl 2,6-difluoro-3-methoxybenzoate ≥95%
Methyl 2,6-difluoro-3-methoxybenzoate
Precaución: for Research Use Only
Formula:
C₉H₈F₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD11520395 CAS Number: 1379300-37-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...