N-(5-Bromo-4-fluoro-2-methylphenyl)acetamide ≥97%

Supplier: AOBChem USA

Warning

32798-1G 32798-25G 32798-5G
76940-326EA 64.72 USD
76940-326 76940-328 76940-330
N-(5-Bromo-4-fluoro-2-methylphenyl)acetamide ≥97%
N-(5-Bromo-4-fluoro-2-methylphenyl)acetamide

Precaución: for Research Use Only

Formula: C₉H₉BrFNO
MW: 246.076 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32070648
CAS Number: 1237518-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR