Methyl 5-bromo-2-hydroxy-3-methylbenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
32730-1G
32730-25G
32730-500MG
32730-5G
76940-312EA
284.33
USD
76940-312
76940-314
76940-316
76940-318
Methyl 5-bromo-2-hydroxy-3-methylbenzoate ≥97%
Methyl 5-bromo-2-hydroxy-3-methylbenzoate
Precaución: for Research Use Only
Formula:
C₉H₉BrO₃ MW: 245.07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD16293433 CAS Number: 40912-71-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...