Methyl 3-chloro-2-methoxypyridine-4-carboxylate ≥97%

Supplier: AOBChem USA

Warning

32601-1G 32601-500MG 32601-5G
76940-270EA 558.04 USD
76940-270 76940-272 76940-274
Methyl 3-chloro-2-methoxypyridine-4-carboxylate ≥97%
Methyl 3-chloro-2-methoxypyridine-4-carboxylate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₃
MW: 201.607 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698151
CAS Number: 1214335-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR