Ethyl 3-chloro-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

32537-25G 32537-5G
76940-260EA 460.44 USD
76940-260 76940-262
Ethyl 3-chloro-4-methylbenzoate ≥97%
Ethyl 3-chloro-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
MW: 198.646 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10566204
CAS Number: 99500-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR