Methyl 3-chloro-5-methoxybenzoate ≥97% research grade

Supplier: AOBChem USA

Warning

32492-25G 32492-5G
76940-250EA 403.15 USD
76940-250 76940-252
Methyl 3-chloro-5-methoxybenzoate ≥97% research grade
Methyl 3-chloro-5-methoxybenzoate
Formula: C₉H₉ClO₃
Storage Temperature: Ambient
MDL Number: MFCD12547160
CAS Number: 96826-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR