6-Fluoro-2-methoxy-3-methylbenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

32489-1G 32489-25G 32489-500MG 32489-5G
76940-234EA 439.22 USD
76940-234 76940-236 76940-238 76940-240
6-Fluoro-2-methoxy-3-methylbenzyl alcohol ≥97%
6-Fluoro-2-methoxy-3-methylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₁FO₂
MW: 170.181 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28519147
CAS Number: 1785476-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR