Methyl-6-fluoropicolinate ≥97%
Supplier: AOBChem USA
|
Warning
|
Synonyms:
Methyl 6-fluoropicolinate
32448-1G
32448-25G
32448-5G
76940-220EA
64.72
USD
76940-220
76940-222
76940-224
Methyl-6-fluoropicolinate ≥97%
Methyl 6-fluoropicolinate
Precaución: for Research Use Only
Formula:
C₇H₆FNO₂ MW: 155.1300 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11110421 CAS Number: 455-71-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...