5-(Hydroxymethyl)isoquinoline ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Isoquinolin-5-ylmethanol, (Isoquinolin-5-yl)methanol

32389-1G 32389-25G 32389-500MG 32389-5G
76940-204EA 177.18 USD
76940-204 76940-206 76940-208 76940-210
5-(Hydroxymethyl)isoquinoline ≥97%
5-(Hydroxymethyl)isoquinoline

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MW: 159.185 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03840599
CAS Number: 76518-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR