6-Chloro-2-methylpyridine-3-boronic acid pinacol ester ≥97%
Supplier: AOBChem USA
|
Warning
|
32300-1G
32300-25G
32300-5G
76940-186EA
62.6
USD
76940-186
76940-188
76940-190
6-Chloro-2-methylpyridine-3-boronic acid pinacol ester ≥97%
6-Chloro-2-methylpyridine-3-boronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₇BClNO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD16996687 CAS Number: 1436866-79-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...