N,N-Dimethylpyridin-3-amine ≥97%

Supplier: AOBChem USA

Warning

32237-25G 32237-5G
76940-158EA 346.92 USD
76940-158 76940-160
N,N-Dimethylpyridin-3-amine ≥97%
N,N-Dimethylpyridin-3-amine
Formula: C₇H₁₀N₂
Storage Temperature: Ambient
MDL Number: MFCD00234349
CAS Number: 18437-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR