Methyl 4-chloro-2-fluoro-3-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

32216-1G 32216-25G 32216-5G
76940-130EA 66.84 USD
76940-130 76940-132 76940-134
Methyl 4-chloro-2-fluoro-3-methoxybenzoate ≥97%
Methyl 4-chloro-2-fluoro-3-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈ClFO₃
MW: 218.609 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD30344686
CAS Number: 1993479-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR