Methyl 3-fluoro-2-methylbenzoate ≥97% research grade

Supplier: AOBChem USA

Warning

32167-1G 32167-25G 32167-500MG 32167-5G
76940-100EA 195.21 USD
76940-100 76940-102 76940-104 76940-106
Methyl 3-fluoro-2-methylbenzoate ≥97% research grade
Methyl 3-fluoro-2-methylbenzoate
Formula: C₉H₉FO₂
MW: 168.165 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06203865
CAS Number: 230301-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR