Methyl 3-fluoro-5-methylbenzoate ≥97% research grade

Supplier: AOBChem USA

Warning

32127-25G 32127-5G
76940-090EA 580.32 USD
76940-090 76940-092
Methyl 3-fluoro-5-methylbenzoate ≥97% research grade
Methyl 3-fluoro-5-methylbenzoate
Formula: C₉H₉FO₂
Storage Temperature: Ambient
MDL Number: MFCD06204167
CAS Number: 660416-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR